![Mithramycin A from Streptomyces plicatus ≥90% (HPLC)](/deepweb/assets/sigmaaldrich/product/structures/301/326/fdf59f27-a69b-42fe-ba80-f5d9f517bf08/640/fdf59f27-a69b-42fe-ba80-f5d9f517bf08.png)
Mithramycin A from Streptomyces plicatus
![Mithramycin A from Streptomyces plicatus ≥90% (HPLC)](/deepweb/assets/sigmaaldrich/product/structures/301/326/fdf59f27-a69b-42fe-ba80-f5d9f517bf08/640/fdf59f27-a69b-42fe-ba80-f5d9f517bf08.png)
Synonym(s):
Aureolic acid, Plicamycin
Empirical Formula (Hill Notation):
C52H76O24
CAS Number:
Molecular Weight:
1085.15
Products
Product Category
Brand
Formula Weight
Purity
Activity Spectrum
Quality Segment
Mode of Action
Showing 1-1 of 1
Compare
Product Number
Description
Pricing