
Malaoxon

Synonym(s):
Diethyl 2-[(dimethoxyphosphoryl)sulfanyl]succinate
Empirical Formula (Hill Notation):
C10H19O7PS
CAS Number:
Molecular Weight:
314.29
Products
Product Category
Brand
Formula Weight
Manufacturer
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2