
Levadopa Related Compound A

Synonym(s):
2,5-Dihydroxy-L-tyrosine, 3-(2,4,5-Trihydroxyphenyl)-L-alanine
Empirical Formula (Hill Notation):
C9H11NO5
CAS Number:
Molecular Weight:
213.19
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing