
Leflunomide for peak identification

Synonym(s):
Leflunomide, 5-Methylisoxazole-4-(4-trifluoromethyl)carboxanilide
Empirical Formula (Hill Notation):
C12H9F3N2O2
CAS Number:
Molecular Weight:
270.21
Products
Formula Weight
Gene Alias
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing