Skip to Content
Merck
L-Aspartic acid-13C4 98 atom % 13C, 95% (CP)

L-Aspartic acid-13C4

L-Aspartic acid-13C4 98 atom % 13C, 95% (CP)
Synonym(s):
(S)-(+)-Aminosuccinic acid-13C4, (S)-Aminobutanedioic acid-13C4, 13C Labeled L-aspartic acid
Linear Formula:
HO213C13CH213CH(NH2)13CO2H
CAS Number:
Molecular Weight:
137.07

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Isotope
Markush Class
Markush Group
Optical Isomer
Showing 1-1 of 1
Compare
Product Number
Description
Pricing