
L-Argininosuccinic acid lithium salt

Synonym(s):
N-{{[(4S)-4-Amino-4-carboxybutyl]amino}iminomethyl}-L-aspartic acid lithium salt, Lithium L-argininosuccinate
Empirical Formula (Hill Notation):
C10H18N4O6 · xLi+
Molecular Weight:
290.27 (free acid basis)
Products
Product Category
Brand
Color
Formula Weight
Manufacturer
Physical Form
Product Line
Purity
Quality Segment
Application
Optical Isomer
Reference Material Type
Special Grade
Showing 1-3 of 3