
Lamivudine for system suitability 1

Synonym(s):
Lamivudine, 2′,3′-Dideoxy-3′-thiacytidine, 3TC, Epivir, Epivir-HBV, Heptovir, Zeffix
Empirical Formula (Hill Notation):
C8H11N3O3S
CAS Number:
Molecular Weight:
229.26
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing