
Ketamine Related Compound A

Synonym(s):
1-[(2-Chlorophenyl)(methylimino)methyl]cyclopentanol
Empirical Formula (Hill Notation):
C13H16ClNO
CAS Number:
Molecular Weight:
237.73
Products
Brand
Formula Weight
Manufacturer
Physical Form
Agency
Format
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2