
Isoorientin

Synonym(s):
Homoorientin, Luteolin 6-C-β-D-glucoside, Luteolin 6-C-glucoside
Empirical Formula (Hill Notation):
C21H20O11
CAS Number:
Molecular Weight:
448.38
Products
Product Category
Brand
Color
Formula Weight
Manufacturer
Physical Form
Purity
Tissue
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing