
Human glucagon

Synonym(s):
Glucagon
Empirical Formula (Hill Notation):
C153H225N43O49S
CAS Number:
Molecular Weight:
3482.75
Products
Formula Weight
Gene Alias
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing