
Gastrodin

Synonym(s):
(2R,3S,4S,5R,6S)-2-Hydroxymethyl-6-(4-hydroxymethyl-phenoxy)-tetrahydro-pyran-3,4,5-triol, 4-β-D-glucopyranosyloxybenzyl alcohol, 4-Hydroxybenzyl alcohol 4-O-β-D-glucoside
Empirical Formula (Hill Notation):
C13H18O7
CAS Number:
Molecular Weight:
286.28
Products
Formula Weight
Manufacturer
Product Line
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing