
Gabapentin impurity D

Synonym(s):
1-[(3-Oxo-2-azaspiro[4.5]dec-2-yl)methyl]cyclohexaneacetic acid
Empirical Formula (Hill Notation):
C18H29NO3
CAS Number:
Molecular Weight:
307.43
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing