
Fluorescein impurity C

Synonym(s):
2-(2,4-Dihydroxybenzoyl)benzoic acid
Empirical Formula (Hill Notation):
C14H10O5
CAS Number:
Molecular Weight:
258.23
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing