
Fenbendazole

Synonym(s):
Methyl 5-(phenylthio)-2-benzimidazolecarbamate, Fenbendazole
Empirical Formula (Hill Notation):
C15H13N3O2S
CAS Number:
Molecular Weight:
299.35
Products
Product Category
Brand
Formula Weight
Manufacturer
Physical Form
Product Line
Purity
Agency
Format
Quality Segment
Application
Markush Class
Markush Group
Package Size
API Family
Reference Material Type
Special Grade
Technique
Showing 1-6 of 6
Compare
Product Number
Description
Pricing