Skip to Content
Merck
Fast Red RC Salt

Fast Red RC Salt

Fast Red RC Salt
Synonym(s):
2-Amino-4-chloroanisole diazotated zinc chloride complex, 5-Chloro-2-methoxybenzenediazonium chloride hemi(zinc chloride) salt, Diazo Fast Red RC
Empirical Formula (Hill Notation):
C7H6Cl2N2O · 0.5ZnCl2
CAS Number:
Molecular Weight:
273.19
EC Number:

Products

Product Category
Brand
Color
Formula Weight
Physical Form
Chemical Composition
Quality Segment
Application
Showing 1-1 of 1
Compare
Product Number
Description
Pricing