
Ethylbenzene solution

Empirical Formula (Hill Notation):
C8H10
CAS Number:
Molecular Weight:
106.17
Products
Product Category
Brand
Formula Weight
Physical Form
Format
Chemical Composition
Quality Segment
Markush Class
Markush Group
Special Grade
Technique
Showing 1-16 of 16
Compare
Product Number
Description
Pricing