![Ethyl 4-ethoxybenzoate certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland](/deepweb/assets/sigmaaldrich/product/structures/300/739/f6a66481-bf74-46a1-984d-6c14a2659a6b/640/f6a66481-bf74-46a1-984d-6c14a2659a6b.png)
Ethyl 4-ethoxybenzoate
![Ethyl 4-ethoxybenzoate certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland](/deepweb/assets/sigmaaldrich/product/structures/300/739/f6a66481-bf74-46a1-984d-6c14a2659a6b/640/f6a66481-bf74-46a1-984d-6c14a2659a6b.png)
Empirical Formula (Hill Notation):
C11H14O3
CAS Number:
Molecular Weight:
194.23
Products
Brand
Formula Weight
Manufacturer
Product Line
Quality Segment
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing