![DMT-dT Phosphoramidite configured for ABI](/deepweb/assets/sigmaaldrich/product/structures/140/215/9cb4d958-3648-49d5-882c-b94976c90ff2/640/9cb4d958-3648-49d5-882c-b94976c90ff2.png)
DMT-dT Phosphoramidite
![DMT-dT Phosphoramidite configured for ABI](/deepweb/assets/sigmaaldrich/product/structures/140/215/9cb4d958-3648-49d5-882c-b94976c90ff2/640/9cb4d958-3648-49d5-882c-b94976c90ff2.png)
Synonym(s):
5′-O-(4,4′-dimethoxytrityl)-2′-deoxythymidine-3′-O-[O-(2-cyanoethyl)-N,N′-diisopropylphosphoramidite], 5′-O-[bis(4-methoxyphenyl)phenylmethyl]-2′-deoxythymidine, 3′-[2-cyanoethyl N,N-bis(1-methylethyl)phosphoramidite], DMT-dT amidite, Thymidine, 5′-O-[bis(4-methoxyphenyl)phenylmethyl]-, 3′-[2-cyanoethyl N,N-bis(1-methylethyl)phosphoramidite]
Empirical Formula (Hill Notation):
C40H49N4O8P
CAS Number:
Molecular Weight:
744.81
Products
Product Category
Brand
Biological Source
Color
Formula Weight
Physical Form
Product Line
Quality Segment
Technique
Showing 1-6 of 6
Compare
Product Number
Description
Pricing