Skip to Content
Merck
(±)-Camphor purum, synthetic, ≥95.0% (GC)

(±)-Camphor

(±)-Camphor purum, synthetic, ≥95.0% (GC)
Synonym(s):
1,7,7-Trimethylbicyclo[2.2.1]heptan-2-one, DL-Camphor, (±)-Camphor
Empirical Formula (Hill Notation):
C10H16O
CAS Number:
Molecular Weight:
152.23
EC Number:

Products

Product Category
Brand
Biological Source
Boiling Point (°C)
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Product Line
Purity
Agency
Quality Segment
Functional Group
Application
Markush Class
Markush Group
Optical Isomer
Reference Material Type
Special Grade