
Carisoprodol

Synonym(s):
2-Methyl-2-propyl-1,3-propanediol carbamate isopropylcarbamate
Empirical Formula (Hill Notation):
C12H24N2O4
CAS Number:
Molecular Weight:
260.33
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing