![Cannabidiol United States Pharmacopeia (USP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/360/583/7344400c-79ad-46f6-ada6-2934c3c8d3f9/640/7344400c-79ad-46f6-ada6-2934c3c8d3f9.png)
Cannabidiol
![Cannabidiol United States Pharmacopeia (USP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/360/583/7344400c-79ad-46f6-ada6-2934c3c8d3f9/640/7344400c-79ad-46f6-ada6-2934c3c8d3f9.png)
Synonym(s):
2-[(1R,6R)-3-Methyl-6-(1-methylethenyl)-2-cyclohexen-1-yl]-5-pentyl-1,3-benzenediol
Empirical Formula (Hill Notation):
C21H30O2
CAS Number:
Molecular Weight:
314.46
Products
Formula Weight
Manufacturer
Physical Form
Product Line
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing