
Calycosin 7-glucoside

Synonym(s):
Calycosin 7-O-β-D-glucopyranoside, 3′,7-Dihydroxy 4′-methoxyisoflavone 7-β-D-glucopyranoside
Empirical Formula (Hill Notation):
C22H22O10
CAS Number:
Molecular Weight:
446.40
Products
Formula Weight
Manufacturer
Physical Form
Product Line
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing