
Bupropion Hydrochloride Related Compound A

Synonym(s):
2-(tert-Butylamino)-4′-chloropropiophenone hydrochloride
Empirical Formula (Hill Notation):
C13H18ClNO · HCl
CAS Number:
Molecular Weight:
276.20
Products
Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2