Skip to Content
Merck
Betamethasone 17-valerate VETRANAL®, analytical standard

Betamethasone 17-valerate

Betamethasone 17-valerate VETRANAL®, analytical standard
Synonym(s):
1,4-Pregnadiene-11β,17α,21-triol-9α-fluoro-16β-methyl-3,20-dione 17-valerate, 9α-Fluoro-16β-methyl-11β,17α,21-trihydroxy-1,4-pregnadiene-3,20-dione 17-valerate, 9α-Fluoro-16β-methylprednisolone 17-valerate, Betnovate
Empirical Formula (Hill Notation):
C27H37FO6
CAS Number:
Molecular Weight:
476.58
EC Number:

Products

Product Category
Brand
Formula Weight
Manufacturer
Physical Form
Product Line
Format
Research Area
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Technique
Showing 1-3 of 3
Compare
Product Number
Description
Pricing