Bacopaside I
Synonym(s):
Pseudojujubogenin 3-O-[α-L-arabinofuranosyl-(1→2)-[6-O-sulfo-β-D-glucopyranosyl-(1→3)]-α-L-arabinopyranoside]
Empirical Formula (Hill Notation):
C46H74O20S
CAS Number:
Molecular Weight:
979.13
Products
Formula Weight
Manufacturer
Physical Form
Product Line
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing