
Arnicolide C

Empirical Formula (Hill Notation):
C19H26O5
CAS Number:
Molecular Weight:
334.41
Products
Product Category
Brand
Biological Source
Formula Weight
Manufacturer
Physical Form
Product Line
Research Area
Application
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing