
apo-Transferrin human

Synonym(s):
Human transferrin, Siderophilin
CAS Number:
EC Number:
Products
Product Category
Brand
Biological Source
Gene Alias
Physical Form
Product Line
Purity
Tissue
Quality Segment
Sterilization Method
Technique
Showing 1-5 of 5
Compare
Product Number
Description
Pricing