![Amphotericin B solution 250 μg/mL in deionized water, 0.1 μm filtered, BioReagent, suitable for cell culture](/deepweb/assets/sigmaaldrich/product/structures/140/115/a0c927b2-145e-4d58-8265-4ecdb3f66477/640/a0c927b2-145e-4d58-8265-4ecdb3f66477.png)
Amphotericin B
![Amphotericin B solution 250 μg/mL in deionized water, 0.1 μm filtered, BioReagent, suitable for cell culture](/deepweb/assets/sigmaaldrich/product/structures/140/115/a0c927b2-145e-4d58-8265-4ecdb3f66477/640/a0c927b2-145e-4d58-8265-4ecdb3f66477.png)
Synonym(s):
Amphotericin B
Empirical Formula (Hill Notation):
C47H73NO17
CAS Number:
Molecular Weight:
924.08
Products
Product Category
Brand
Color
Formula Weight
Manufacturer
Physical Form
Product Line
Activity Spectrum
Agency
Format
Quality Segment
Application
Mode of Action
Package Size
API Family
Reference Material Type
Special Grade
Sterilization Method
Technique
Showing 1-4 of 4
Compare
Product Number
Description
Pricing