
Amoxapine Related Compound D

Synonym(s):
2-Chlorodibenz[b,f][1,4]oxazepin-11(10H)-one, 2-Chlorodibenzo[b,f][1,4]oxazepin-11-one
Empirical Formula (Hill Notation):
C13H8ClNO2
CAS Number:
Molecular Weight:
245.66
Products
Brand
Formula Weight
Manufacturer
Physical Form
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing