Skip to Content
Merck
Amastatin hydrochloride hydrate ≥97% (HPLC)

Amastatin hydrochloride hydrate

Amastatin hydrochloride hydrate ≥97% (HPLC)
Synonym(s):
(2S,3R)-3-Amino-2-hydroxy-5-methylhexanoyl-Val-Val-Asp hydrochloride hydrate
Empirical Formula (Hill Notation):
C21H38N4O8 · HCl · xH2O
CAS Number:
Molecular Weight:
511.01 (anhydrous basis)

Products

Product Category
Brand
Biological Source
Formula Weight
Physical Form
Purity
Activity Spectrum
Quality Segment
Mode of Action
Showing 1-1 of 1
Compare
Product Number
Description
Pricing