Skip to Content
Merck
Allyl tetraisopropylphosphorodiamidite 95%

Allyl tetraisopropylphosphorodiamidite

Allyl tetraisopropylphosphorodiamidite 95%
Linear Formula:
[[(CH3)2CH]2N]2POCH2CH=CH2
CAS Number:
Molecular Weight:
288.41

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing