
Aciclovir for impurity G identification

Synonym(s):
Acycloguanosine, 9-[(2-Hydroxyethoxy)methyl]guanine, Acyclovir
Empirical Formula (Hill Notation):
C8H11N5O3
CAS Number:
Molecular Weight:
225.20
Products
Formula Weight
Manufacturer
Application
API Family
Reference Material Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing