
Acebutolol impurity C

Synonym(s):
N-(3-Acetyl-4-hydroxyphenyl)butanamide, 2-Acetyl 4-butyramidophenol
Linear Formula:
CH3CH2CH2CONHC6H3(OH)COCH3
CAS Number:
Molecular Weight:
221.25
Products
Formula Weight
Manufacturer
Melting Point (°C)
Format
Application
Markush Class
Markush Group
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing