Skip to Content
Merck
5-Fluoroorotic acid hydrate 95%

5-Fluoroorotic acid hydrate

5-Fluoroorotic acid hydrate 95%
Empirical Formula (Hill Notation):
C5H3FN2O4 · xH2O
Molecular Weight:
174.09 (anhydrous basis)

Products

Brand
Formula Weight
Physical Form
Purity
Quality Segment
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing