![5,10,15,20-Tetrakis(pentafluorophenyl)porphyrin ≥90.0% (HPLC)](/deepweb/assets/sigmaaldrich/product/structures/109/988/c1e7cb39-b4de-4207-bb27-8002080de6d6/640/c1e7cb39-b4de-4207-bb27-8002080de6d6.png)
5,10,15,20-Tetrakis(pentafluorophenyl)porphyrin
![5,10,15,20-Tetrakis(pentafluorophenyl)porphyrin ≥90.0% (HPLC)](/deepweb/assets/sigmaaldrich/product/structures/109/988/c1e7cb39-b4de-4207-bb27-8002080de6d6/640/c1e7cb39-b4de-4207-bb27-8002080de6d6.png)
Empirical Formula (Hill Notation):
C44H10F20N4
CAS Number:
Molecular Weight:
974.55
Products
Brand
Formula Weight
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing