Skip to Content
Merck
3-Deoxy-2-keto-6-phosphogluconic acid lithium salt ≥95% (TLC)

3-Deoxy-2-keto-6-phosphogluconic acid lithium salt

3-Deoxy-2-keto-6-phosphogluconic acid lithium salt ≥95% (TLC)
Synonym(s):
(4S,5R)-4,5-Dihydroxy-2-oxo-6-phosphonooxyhexanoic acid lithium salt, 2-Dehydro-3-deoxy-6-phospho-D-gluconate lithium salt, 2-Keto-3-deoxy-6-phosphogluconate lithium salt, 3-Deoxy-2-keto-D-gluconic acid 6-phosphate lithium salt, 3-Deoxy-D-erythro-hexulosonic acid 6-phosphate lithium salt, KDPG
Empirical Formula (Hill Notation):
C6H11O9P · xLi+
CAS Number:
Molecular Weight:
258.12 (free acid basis)

Products

Product Category
Brand
Color
Formula Weight
Physical Form
Purity
Optical Isomer
Showing 1-1 of 1
Compare
Product Number
Description
Pricing