Skip to Content
Merck
3-Chloro-2-hydroxy-1-propanesulfonic acid sodium salt hydrate 95%

3-Chloro-2-hydroxy-1-propanesulfonic acid sodium salt hydrate

3-Chloro-2-hydroxy-1-propanesulfonic acid sodium salt hydrate 95%
Linear Formula:
ClCH2CH(OH)CH2SO3Na · xH2O
CAS Number:
Molecular Weight:
196.59 (anhydrous basis)
EC Number:

Products

Product Category
Formula Weight
Melting Point (°C)
Purity
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing