Skip to Content
Merck
2-Chloropropionic acid 92%

2-Chloropropionic acid

2-Chloropropionic acid 92%
Synonym(s):
(±)-2-Chloropropionic acid, 2-Chloropropionic acid
Linear Formula:
CH3CHClCOOH
CAS Number:
Molecular Weight:
108.52

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Showing 1-2 of 2
Compare
Product Number
Description
Pricing