
2-Azahypoxanthine

Synonym(s):
3,7-Dihydro-4H-imidazo[4,5-d]-1,2,3-triazin-4-one, 2-Azahypoxanthine, NSC 22419
Empirical Formula (Hill Notation):
C4H3N5O
CAS Number:
Molecular Weight:
137.10
Products
Formula Weight
Manufacturer
Physical Form
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing