ASTM50
ASTM Colour Reference Standard ASTM D 6045, D1500 (ASTM colour 5)
ASTM D 6045, D1500 (ASTM colour 5)
Synonym(s):
ASTM Colour Reference Standard
About This Item
Recommended Products
grade
certified reference material
Agency
according to ASTM® D 6045
according to ASTM® D1500
shelf life
limited shelf life, expiry date on the label
manufacturer/tradename
Paragon Scientific Ltd
application(s)
cleaning products
coatings
cosmetics
environmental
flavors and fragrances
food and beverages
glass & ceramic
personal care
pharmaceutical
format
mixture
Looking for similar products? Visit Product Comparison Guide
General description
Application
Features and Benefits
- Liquid colour standard compliant to ASTM D 6045 and D1500
- Certified as per the dual UKAS accreditation to BS EN ISO / IEC 17025 and ISO 17034
- Traceable to internationally recognized standards with high accuracy
- Delivered in 500-mL tamper-evident bottles with safety data sheet
- High-stability material with 18-months shelf life
Legal Information
Storage Class Code
10 - Combustible liquids
WGK
WGK 1
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service