Key Documents
Z688533
Microscope slide-grids, disposable
use with dry or suspended specimens
About This Item
UNSPSC Code:
41122600
NACRES:
MA.01
material
polyester
packaging
pack of 40 ea
manufacturer/tradename
Diversified Biotech GRID-1000
W × L
20 mm × 20 mm
General description
Convenient- Easy-to adhere stickers having side-by-side twin grids (20 mm x 20 mm each with 1 mm line spacing) for microscope slides. Count or measure cells and estimate the size or length of microscopic or semi-micro objects
Versatile- Compatible with different styles of microscopes (conventional, inverted or dissecting). Virtually all cells and specimens placed on top of Slide-Grids can be counted and/or sized.
Dry or Wet- Use with dry specimens or suspended specimens such as cells held in buffer under cover slips. Separate cover slips can be placed over each Slide-Grid.
Chemically Inert- Made of polyester with an acrylic adhesive. Slide-Grids are inert and stable under most laboratory conditions.
Cover slips not included
Choose from one of the most recent versions:
Certificates of Analysis (COA)