U4510
UNISETTE™ Tissue Processing/Embedding Cassettes
green
Synonym(s):
UNISETTE™ Tissue Processing/Embedding Cassettes
Sign Into View Organizational & Contract Pricing
All Photos(3)
About This Item
UNSPSC Code:
41116124
NACRES:
NB.17
Recommended Products
color
green
application(s)
hematology
histology
storage temp.
room temp
Looking for similar products? Visit Product Comparison Guide
General description
Histology processing/embedding cassettes made of acetal polymer. These cassettes keep specimens safely submerged in liquid and are resistant to histological solvents. The one-piece lid snaps apart and conveniently locks into the cassette base, ensuring specimen safety. A line of cassette colors are available to fit your needs and preference. A large 30° anterior writing area is available for your convenience. Not suitable for automated printers.
Application
Compatible with Leica and Sakura printers.
Quantity
Case contains 3 dispenser boxes of 500 cassettes.
Legal Information
Unisette is a trademark of Simport
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service