Key Documents
EP0030129601
Eppendorf twin.tec® PCR Plates 96
Skirted, Forensic DNA Grade, working volume 150 μL, colorless, pack of 10 (individually wrapped)
About This Item
UNSPSC Code:
41106300
NACRES:
NB.22
material
colorless
colorless wells
well size
96
feature
skirt
Forensic DNA Grade
Skirt
Skirted
packaging
pack of 10 (individually wrapped)
manufacturer/tradename
Eppendorf® 0030129601 (Global)
technique(s)
PCR: suitable
size
96 well
working volume
150 μL
Application
PCR
Features and Benefits
  • Maximum recovery rates for improved assay results
  • Available with barcode (upon request)
  • Ideal for quantitative real-time PCR as well
  • Exceptionally solid and torque-resistant polycarbonate frame
  • OptiTrack® matrix for faster sample identification and fewer pipetting errors
  • Raised well rims for effective sealing, also reduces risk of cross-contamination
  • Batch-tested and certified free of DNA, DNAase, RNase and PCR inhibitors (PCR clean)
Legal Information
Eppendorf is a registered trademark of Eppendorf AG
OptiTrack is a registered trademark of Eppendorf AG
twin.tec is a registered trademark of Eppendorf AG