Skip to Content
Merck
All Photos(1)

Key Documents

ERMAC213

PAHs in toluene

ERM®, certified reference material

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

Agency

ERM®

manufacturer/tradename

JRC

format

multi-component solution

storage temp.

−20°C

Application

PAHs in toluene has been used as a certified reference material for the quantification of semi-volatile organic compounds in air samples using gas chromatography-mass spectrometry analysis (GC–MS).

Analysis Note

For more information please see:
ERMAC213

Legal Information

ERM is a registered trademark of European Commission

Signal Word

Danger

Hazard Classifications

Aquatic Chronic 3 - Asp. Tox. 1 - Flam. Liq. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 2 Inhalation - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

39.9 °F - closed cup

Flash Point(C)

4.4 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Optimisation steps of an innovative air sampling method for semi volatile organic compounds
Lazarov B, et al.
Atmospheric Environment, 79, 780-786 (2013)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service