Skip to Content
Merck
All Photos(1)

Key Documents

71333

Sigma-Aldrich

pIEx-6 DNA - Novagen

Novagen′s pIEx-6 vector is designed to allow rapid, high-level protein expression in insect cells without the time-consuming process of creating a recombinant baculovirus.

Synonym(s):

InsectDirect System- pIEx-6 DNA

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106612
NACRES:
NA.51

Quality Level

form

solid

manufacturer/tradename

Novagen®

storage condition

OK to freeze

shipped in

wet ice

storage temp.

−70°C

Warning

Toxicity: Standard Handling (A)

Legal Information

INSECTDIRECT is a trademark of Merck KGaA, Darmstadt, Germany
NOVAGEN is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service