1.11143
Chlorine and pH Test Kit Refill pack, colorimetric
for use with MQUANT®
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
product name
Chlorine and pH Test Refill pack Reagent pH-1, for use with MQUANT®
feature
refill
Quality Level
specific analyte(s)
chlorine
compatibility
for use with MQUANT®
detection method
colorimetric
storage temp.
15-25°C
General description
When the reagents of a MQuant® test (formally MColortest®) are used up, the kit can be replenished by ordering the appropriate refill pack. Refill packs are available for a range of test kits, making it a particularly economical and ecological system.
This refill pack contains all the reagents for the MQuant® test kit 1.11174 for the colometric evaluation of pH.
This refill pack contains all the reagents for the MQuant® test kit 1.11174 for the colometric evaluation of pH.
for 111174 MQuant®
Legal Information
MCOLORTEST is a registered trademark of Merck KGaA, Darmstadt, Germany
MQUANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service