air monitoring cosmetics food and beverages pharmaceutical
compatibility
for use with MAS-100 Eco® for use with MAS-100 NT® for use with MAS-100 VF®
storage temp.
no temp limit
General description
Tube-Adapter for portable MAS-100® viable air sampler is made of anodized aluminum and fits all perforated lids of the portable MAS-100® product line. The tube adapter along with suitable tubing and the MAS-100® sampler allows microbial air sampling at sampling points that are difficult to access such as floor outlets and air ducts. Due to its geometric design it ensures isokinetic collection with the MAS-100® air samplers.
Application
Tube-Adapter for portable MAS-100® air sampler devices are extensively used in Pharmaceutical and Personal care industries as well as in Food, Beverage and Environmental applications.
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.