Skip to Content
Merck
All Photos(2)

Key Documents

713376

Sigma-Aldrich

tert-Butyl (phenylmethylene)carbamate

≥98.0%

Synonym(s):

N-(tert-Butoxycarbonyl)benzylideneamine, N-Boc-benzaldimine, N-[Phenylmethlene]-carbamic acid tert-butyl ester, tert-Butyl N-benzylidenecarbamate, Benzaldehyde N-(tert-butoxycarbonyl)imine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H15NO2
CAS Number:
Molecular Weight:
205.25
Beilstein:
6324080
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

≥98.0% (GC)
≥98.0%

form

liquid

storage temp.

2-8°C

SMILES string

CC(C)(C)OC(=O)\N=C\c1ccccc1

InChI

1S/C12H15NO2/c1-12(2,3)15-11(14)13-9-10-7-5-4-6-8-10/h4-9H,1-3H3/b13-9+

InChI key

XFRQMBFCAKTYIV-UKTHLTGXSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service