Skip to Content
Merck
All Photos(1)

Key Documents

442801

Sigma-Aldrich

2-Hydroxy-5-(trifluoromethyl)pyridine

97%

Synonym(s):

5-(Trifluoromethyl)-2-pyridinol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H4F3NO
CAS Number:
Molecular Weight:
163.10
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

mp

145-149 °C (lit.)

SMILES string

Oc1ccc(cn1)C(F)(F)F

InChI

1S/C6H4F3NO/c7-6(8,9)4-1-2-5(11)10-3-4/h1-3H,(H,10,11)

InChI key

BYRJSCNPUHYZQE-UHFFFAOYSA-N

General description

2-Hydroxy-5-(trifluoromethyl)pyridine, a pyridine derivative, is a heterocyclic building block. They have useful applications in catalysis, drug design, molecular recognition, and natural product synthesis.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nora Badawi et al.
Environmental science & technology, 49(15), 8995-9003 (2015-07-07)
The herbicide fluazifop-P-butyl (FPB) is used against grasses in agricultural crops such as potato, oilseed rape, and sugar beet. Limited information is available in scientific literature on its environmental fate, therefore extensive monitoring at two agricultural test fields was combined
N J Bailey et al.
Journal of agricultural and food chemistry, 48(1), 42-46 (2000-01-19)
Directly coupled HPLC-NMR-MS was used to characterize two major metabolites of 5-trifluoromethylpyridone (2-hydroxy-5-trifluoromethylpyridine), a model compound for herbicides, after it had been dosed into hydroponically grown maize plants. The combination of NMR and MS data allowed the identification of both

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service