Merck
  • Tributyltin exposure increases mortality of nodavirus infected Japanese medaka Oryzias latipes larvae.
Tributyltin exposure increases mortality of nodavirus infected Japanese medaka Oryzias latipes larvae.
Marine pollution bulletin (2017-02-23)
Shin-Ichi Kitamura, Masaki Akizuki, Jun-Young Song, Kei Nakayama
ABSTRACT
We investigated the effect of combined exposure to nodavirus infection and TBT on medaka (Oryzias latipes). Medaka larvae were infected by immersion in medium containing nodavirus at titers of 10
Sigma-Aldrich
Tributyltin chloride, 96%